Learn More
Labconco™ Protector™ Acid and Standard Storage Cabinets Shelf Kits
Shelf Kits provide additional storage space in Labconco Protector Standard and Acid Base Cabinets.
Supplier:  Labconco™ 9919000
 View more versions of this product
Catalog No. 1630544
Only null left
Description
Description
Interior Shelf Kit provides additional storage space inside Labconco Protector Acid Storage Cabinets. Includes powder-coated steel shelf, acid-resistant PVC tray, and mounting hardware.
Specifications
Chemical Identifiers
Shelf Kit
Specifications
Protector Acid Storage Cabinet
121.92 cm
48 in.
Shelf Kit
Epoxy Coated Steel shelf, Acid-resistant Polyethylene tray and mounting hardware
Product Suggestions
Product Suggestions