Learn More
Labconco™ Pressure Relief Valve Vent Kits for Precise™ Glove Boxes
Catalog No. 16108430
Change view
Click to view available options
For Use With (Equipment):
Precise Basic Glove Box
Precise Controlled Atmosphere Glove Box
Product Type:
Carbon-filtered Pressure Relief Valve Vent Kit
HEPA-filtered Pressure Relief Valve Vent Kit
Pressure Relief Valve Vent Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
16108430 Precise Basic Glove Box Pressure Relief Valve Vent Kit
16108421 Precise Controlled Atmosphere Glove Box HEPA-filtered Pressure Relief Valve Vent Kit
16108423 Precise Controlled Atmosphere Glove Box Carbon-filtered Pressure Relief Valve Vent Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16108430 Supplier Labconco™ Supplier No. 5242700
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Accessories for Precise Glove Boxes
Pressure Relief Vent Kits provide the components necessary to exhaust the pressure relief valve during overpressurization. Require 5230600 Pressure Relief Valve (included with Precise Controlled Atmosphere Glove Boxes) and outside ducting. Include 3.9 in. OD x 3.8 in. ID thimble connection, gasket, two elbows, four clamps and 13 in. long tubing. Ductwork for outside venting is not included.
Specifications
For Use With (Equipment) Precise Basic Glove Box
Weight (English) 5 lb.
Weight (Metric) 2.3 Kg
Product Type Pressure Relief Valve Vent Kit