Learn More
Labconco™ Exterior Monitor Tubing Kits
Catalog No. 16200481
Change view
Click to view available options
Product Type:
Exterior Moisture Monitor Tubing Kit
Exterior Oxygen Monitor Tubing Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16200481 Exterior Moisture Monitor Tubing Kit
16200480 Exterior Oxygen Monitor Tubing Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16200481 Supplier Labconco™ Supplier No. 5442500
Tubing Kits connect Oxygen or Moisture monitor to glove box
Exterior Monitor Tubing Kit provides attachment tubing to allow placement of Oxygen or Moisture Monitors outside the glove box. For use with Labconco Protector and Precise Glove Boxes. For use with Moisture Monitor 5244900 amd 5244901.
Specifications
Weight (English) 2 lb.
Weight (Metric) 5 Kg
Product Type Exterior Moisture Monitor Tubing Kit