Learn More
Labconco™ Exterior Monitor Tubing Kits
Catalog No. 16200480
Change view
Click to view available options
Product Type:
Exterior Moisture Monitor Tubing Kit
Exterior Oxygen Monitor Tubing Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
16200480 Exterior Oxygen Monitor Tubing Kit
16200481 Exterior Moisture Monitor Tubing Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16200480 Supplier Labconco™ Supplier No. 5442400
Tubing Kits connect Oxygen or Moisture monitor to glove box
Exterior Monitor Tubing Kit provides attachment tubing to allow placement of Oxygen or Moisture Monitors outside the glove box. For use with Labconco Protector and Precise Glove Boxes. For use with Oxygen Monitor 5244500 and 5244501.
Specifications
Weight (English) 2 lb.
Weight (Metric) 5 Kg
Product Type Exterior Oxygen Monitor Tubing Kit