Learn More
Labconco™ Exterior Glove Port Covers
Description
Includes
Two clamps
May be placed over glove ports with or without gloves attached to facilitate purge/fill operations and preserve inert atmosphere
- Molded neoprene construction
- For use with all glove boxes
Specifications
Specifications
| For Use With (Equipment) | All Glove Box |
| Weight (English) | 3.5 lb. |
| Weight (Metric) | 1.6 Kg |
| Product Type | Cover |