Learn More
Labconco™ Duplexes for Basic Hoods
For Labconco Basic 47 and 70 Fume Hoods
Supplier: Labconco™ 4844400
Description
Duplex Electrical Receptacle Kits for use with Labconco Basic 47 and Basic 70 Fume Hoods. Each kit includes duplex electrical receptacle, junction box and receptacle cover plate. For mounting on the lower right side fixture panel. Electrical wiring is not included.Chemical Identifiers
| Basic™ Hood | |
| 20 A | |
| 115 V | |
| Mounting lower right-side panel of Basic™ hood |
Specifications
| Basic™ Hood | |
| Standard Duplex Kit | |
| Mounting lower right-side panel of Basic™ hood |
| 20 A | |
| 115 V |