Learn More
Labconco™ Ceiling Enclosure Kits for Protector™Premier™, XL and XStream™ Laboratory Hoods
Ceiling enclosures extend top of hood to the ceiling to hide exposed ductwork, plumbing and wiring
Supplier:  Labconco™ 9414811
 View more versions of this product
Catalog No. 10-379-222
Only null left
Explore available promotions
Description
Description
  • Fit any 3 - 8 ft. Protector Premier, XL or XStream Laboratory Hood (based on appropriate hood depth and nominal width)
  • Each kit includes three ceiling enclosure panels — one for front of hood and one for each side of the hood
  • Panels are constructed of white, powder-coated steel
Note: The height range should match the height between the top of the hood and the building ceiling. Refer to the product specifications to help determine the appropriate Ceiling Enclosure Kit for your hood.
Specifications
Specifications
Protector Premier Hood
96 in.
243.8 cm
14 to 18.6 in.
95.8 cm
14 to 18.6 in.
35.56 to 47.24 cm
Protector™ Premier™, XStream™, XL™ fume hood
35.56 to 47.24 cm
37.7 in.
Product Suggestions
Product Suggestions