Learn More
Labconco™ Ceiling Enclosure Kits for Protector™Premier™, XL and XStream™ Laboratory Hoods
Ceiling enclosures extend top of hood to the ceiling to hide exposed ductwork, plumbing and wiring
Supplier:  Labconco™ 9414602
 Afficher plus de versions de ce produit
Numéro de catalogue. 10-379-214
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Fit any 3 - 8 ft. Protector Premier, XL or XStream Laboratory Hood (based on appropriate hood depth and nominal width)
  • Each kit includes three ceiling enclosure panels — one for front of hood and one for each side of the hood
  • Panels are constructed of white, powder-coated steel
Note: The height range should match the height between the top of the hood and the building ceiling. Refer to the product specifications to help determine the appropriate Ceiling Enclosure Kit for your hood.
Spécifications
Spécifications
Protector Premier Hood
72 in.
182.8 cm
18.6 to 24.4 in.
80.52 cm
18.6 to 24.4 in.
47.24 to 61.97 cm
Protector™ Premier™, XStream™, XL™ fume hood
47.24 to 61.97 cm
31.7 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS