Learn More
Labconco™ Ceiling Enclosure Kits for Protector™Premier™, XL and XStream™ Laboratory Hoods
Ceiling enclosures extend top of hood to the ceiling to hide exposed ductwork, plumbing and wiring
Supplier: Labconco™ 9414512
Description
- Fit any 3 - 8 ft. Protector Premier, XL or XStream Laboratory Hood (based on appropriate hood depth and nominal width)
- Each kit includes three ceiling enclosure panels — one for front of hood and one for each side of the hood
- Panels are constructed of white, powder-coated steel
Specifications
| Protector Premier Hood | |
| 60 in. | |
| 152 cm | |
| 18.6 to 24.4 in. | |
| 95.8 cm |
| 18.6 to 24.4 in. | |
| 47.24 to 61.97 cm | |
| Protector™ Premier™, XStream™, XL™ fume hood | |
| 47.24 to 61.97 cm | |
| 37.7 in. |