Learn More
Labconco™ Atmospure Tubing Kit
Catalog No. 16200470
Change view
Click to view available options
For Use With (Equipment):
Double Width Glove Box
Single Width Glove Box
Product Type:
Atmospure Flex Tubing Kit, Double
Atmospure Flex Tubing Kit, Single
Atmospure Hard Tubing Kit, Single
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
16200470 Single Width Glove Box Atmospure Flex Tubing Kit, Single
16200471 Double Width Glove Box Atmospure Flex Tubing Kit, Double
16200472 Single Width Glove Box Atmospure Hard Tubing Kit, Single
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16200470 Supplier Labconco™ Supplier No. 5441800
Tubing Kits connect AtmosPure Re-Gen Gas Purifier to glove box
AtmosPure Circulation Tubing Kit includes the plumbing to make the connection from the Labconco AtmosPure Re-Gen Gas Purifier to the glove box. Includes 3/4 in. ID neoprene-lined flexible hose. For use with all single-width glove boxes.
Specifications
For Use With (Equipment) Single Width Glove Box
Weight (English) 20 lb.
Weight (Metric) 9.1 Kg
Product Type Atmospure Flex Tubing Kit, Single