Learn More
Labconco™ Anti-Static Ionizer Fan
Catalog No. 16108403
Change view
Click to view available options
Voltage:
100/115 V
230 V
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
16108403 100/115 V
16108404 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16108403 Supplier Labconco™ Supplier No. 5234400
Circulate ionized air inside Labconco Glove Boxes.
Anti-Static Ionizer Fan circulates ionized air inside Labconco Protector and Precise Glove Boxes and XPert Weigh Boxes to reduce static charge. Fan requires installation.
Specifications
For Use With (Equipment) Protector Combination Glove Box
Weight (English) 3.5 lb.
Weight (Metric) 1.6 Kg
Voltage 100/115 V
For Use With (Application) Circulates ionized air inside Glove Box to reduce Static Charge