missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-2 Ab-5 Mouse Monoclonal Antibody, Epredia™
Description
TIMP-1 and TIMP-2 are inhibitory enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-2 shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-2 or gelatinase A). It also has affinity for the active form of 92kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.
Host Species: Mouse
Clone: 3A4
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Rabbit, Cow and Guinea Ppig. Others not known.
Epitope: N-terminal
Immunogen: A synthetic peptide from the N-terminal region of human TIMP-2 protein
Molecular Weight: 21kDa
Positive Control: Conditioned, serum-free medium from (TPA-treated) HFL-1 cells; Pancreatic or colon CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
- Western Blotting
- Immunoprecipitation (Not suitable)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | TIMP-2 Ab-5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 3A4 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | A synthetic peptide from the N-terminal region of human TIMP-2 protein |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |