missing translation for 'onlineSavingsMsg'
Learn More

Progesterone Receptor Ab-6 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Progesterone Receptor Ab-6, Mouse Monoclonal Antibody.

Supplier:  Thermo Fisher Scientific MS196PABX

 View more versions of this product

Catalog No. MS196PABX

Only null left

Description

Description

Human PgR exists in two forms: 116kDa (B-form) and 81kDa (A-form). It acts as ligand activated transcription factor to regulate expression of the target genes. Null mutation in PgR gene leads to pleiotrophic reproductive abnormalities.

Host Species: Mouse

Clone: hPRa 6

Isotype: IgG2b

Species Reactivity: Human, Cow, Rabbit, and Chicken. Does not react with monkey. Others not known.

Epitope: Not determined

Immunogen: Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice

Molecular Weight: 116kDa (triplet, B-form)

Positive Control: T47D cells

Cellular Localization: Nuclear Ab-6 preferentially reacts with B-form of progesterone receptor.

Recommended for:

  • Western Blotting (Not optimum)
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Sedimentation in Sucrose Gradient

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Progesterone Receptor Ab-6
Monoclonal
1mg/mL
Mouse
200 μL
Cancer and Tumor Biology
Bovine, Chicken, Human, Rabbit
IgG2b
Immunofluorescence, Immunoprecipitation
hPRa 6
Unconjugated
Partially purified PgR from a human endometrial carcinoma (EnCa 101) grown in athymic mice
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions