missing translation for 'onlineSavingsMsg'
Learn More

MHC II (HLA-DR) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ MHC II (HLA-DR) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Supplier:  Thermo Fisher Scientific MS133PABX

 View more versions of this product

Catalog No. MS133PABX


Only null left

Description

Description

HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of an α chain (36 kDa) and a β chain (27 kDa). It is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They therefore have a critical role in the initiation of the immune response.

Host Species: Mouse

Clone: LN3

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Activated human peripheral blood mononuclear cells

Molecular Weight: 36kDa (α-chain) and 27kDa (β-chain)

Positive Control: Tonsil/lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Chemical Identifiers

MHC II (HLA-DR) Ab-1
Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Hematopoietic
Activated human peripheral blood mononuclear cells
Primary
LN3
Monoclonal
Concentrated
RUO
Human
IgG2b

Specifications

MHC II (HLA-DR) Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Hematopoietic
Human
IgG2b
Flow Cytometry, Immunohistochemistry (Paraffin)
LN3
Unconjugated
Activated human peripheral blood mononuclear cells
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions