Learn More
Heat Shock Protein 70 (HSP70) Ab-2 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation and flow cytometry experiments.
Supplier: Thermo Fisher Scientific MS482P1ABX
Description
Heat shock proteins (HSP's) or stress response proteins (SRP's) are synthesized in variety of environmental and pathophysiological stressful conditions. Many HSP's are involved in processes such as protein denaturation-renaturation, folding-unfolding, transport-translocation, activation-inactivation, and secretion. HSP70 is found to be associated with steroid receptors, actin, p53, polyoma T antigen, nucleotides, and other unknown proteins. Also, HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs, and other damaging conditions.Host Species: Mouse
Clone: W27
Isotype: IgG2a
Species Reactivity: Human, Monkey and Cow. Others not known.
Epitope: Not determined
Immunogen: HSP70 protein from HeLa cells
Molecular Weight: 73 kDa (constitutive), 72 kDa (inducible)
Positive Control: BT474 or HeLa cells, Breast CA
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Chemical Identifiers
| Heat Shock Protein 70 (HSP70) Ab-2 | |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Mouse | |
| Stress Response | |
| HSP70 protein from HeLa cells | |
| Primary |
| W27 | |
| Monoclonal | |
| RUO | |
| Bovine, Human, Non-human Primate | |
| IgG2a |
Specifications
| Heat Shock Protein 70 (HSP70) Ab-2 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μL | |
| Stress Response | |
| Bovine, Human, Non-human Primate | |
| IgG2a |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| W27 | |
| Unconjugated | |
| HSP70 protein from HeLa cells | |
| RUO | |
| Primary | |
| Concentrated |