Learn More
Growth Hormone, Human/hGH, Ab-3 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry and western blotting experiments.
Supplier: Thermo Fisher Scientific MS1328P1
Description
hGH is produced by the somatotroph cells in the pituitary. hGH acts through binding to membrane receptors that belong to the cytokine receptor superfamily. Ligand binding induces receptor dimerization and activation of the receptor-associated kinase: JAK2; this results in phosphorylation of the kinase itself, of the receptor, and of many cellular proteins including the Stat proteins as well as adaptors leading to the activation of the Ras/MAP kinase pathway and of the PI-3 kinase pathway.Host Species: Mouse
Clone: GH01/#2 + GH02/#7
Isotype: IgG2a/κ + IgG2a/κ
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant human growth hormone
Molecular Weight: ∽22 kDa
Positive Control: Recombinant human growth hormone, Human anterior pituitary
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Note:
USA: RUO; Int'l: RUO
Chemical Identifiers
| Growth Hormone, Human/hGH Ab-3 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| IgG2a κ |
| GH01 / #2 + GH02 / #7 | |
| Monoclonal | |
| Mouse | |
| Steroids Hormones and Growth Factors | |
| Purified recombinant human growth hormone | |
| Primary |
Specifications
| Growth Hormone, Human/hGH Ab-3 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Steroids Hormones and Growth Factors | |
| Human | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| GH01 / #2 + GH02 / #7 | |
| Unconjugated | |
| Purified recombinant human growth hormone | |
| RUO | |
| Primary | |
| Concentrated |