missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdk4 Ab-1 Mouse Monoclonal Antibody, Epredia™
Description
A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.
Host Species: Mouse
Clone: DCS-35
Isotype: IgG1/κ
Species Reactivity: Human, Mouse, Rat and Pig. Others not known.
Epitope: aa 1-20
Immunogen: Purified recombinant Cdk4 protein
Molecular Weight: 34 kDa
Positive Control: NIH 3T3, MAD109, LS174T, or MCF-7 cells
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native only)
- Kinase Assay
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Cdk4 Ab-1 |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | DCS-35 |
| Concentration | 200μg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Immunogen | Purified recombinant cdk4 protein |
| Quantity | 500 μL |
| Regulatory Status | RUO |
| Show More |