missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody

Catalog No. MS1096S1
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1096S1 500 μL Concentrated
MS1096S0 100 μL Concentrated
MS1096S 1 mL Concentrated
MS1096R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1096S1 Supplier Epredia™ Supplier No. MS1096S1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ensure accurate, reproducible results in immunohistochemistry applications with Epredia™ CD71/Transferrin Receptor Ab-3, Mouse Monoclonal Antibody.

Cell surface transferrin receptor (CD71) is a disulfide-bonded homodimeric glycoprotein. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. Transferrin receptor (CD71) has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.

Host Species: Mouse

Clone: 10F11

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Molecular Weight: ∽90-95 kDa

Positive Control: Placenta

Cellular Localization: Granular cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD71/Transferrin Receptor Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10F11
Conjugate Unconjugated
Host Species Mouse
Purification Method Tonsil
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2b
Show More Show Less