missing translation for 'onlineSavingsMsg'
Learn More

CD35/CR1 (Follicular Dendritic Cell Marker) Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.

Supplier:  Thermo Fisher Scientific MS336R7

 View more versions of this product

Catalog No. MS336R7

Only null left
Explore available promotions

Description

Description

Complement receptor 1 (CR1, CD35) is a cell membrane-bound, monomeric glycoprotein that is found on numerous cell types of primates including erythrocytes, lymphocytes, and phagocytes. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules.

Host Species: Mouse

Clone: E11

Isotype: IgG1

Species Reactivity: Human, Rhesus Monkey and Baboon. Others not tested.

Epitope: Distal from the C3b-binding site

Immunogen: Intact human monocytes

Molecular Weight: 210-220 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD35
Monoclonal
Mouse
7 mL
Hematopoietic
Human, Non-human Primate
IgG1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
E11
Unconjugated
Intact human monocytes
RUO
Primary
Prediluted
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions