missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum, 20 gal.
GREENER_CHOICE

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

Supplier:  Eagle™ 1623MB

 View more versions of this product

Catalog No. 19338438

Only null left

Description

Description

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids
Specifications

Chemical Identifiers

Drum
20 gal.
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Open
75.9 cm
Yes
75.8 L
HDPE
29.9 in.
10.25 lb.

Specifications

Drum
Metal Lever-lock
75.8 L
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Open
75.9 cm
Yes
20 gal.
Blue
HDPE
29.9 in.
10.25 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions