missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Lab Pack Open Head Poly Drum, 20 gal.
GREENER_CHOICE

Catalog No. p-7222716
Change view
Click to view available options
Color:
Black
Blue
Yellow
Closure Type:
Metal Lever-lock
Plastic Lever-lock
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
19338436 Black Plastic Lever-lock
19338435 Blue Plastic Lever-lock
19338434 Yellow Plastic Lever-lock
19338439 Black Metal Lever-lock
19338438 Blue Metal Lever-lock
19338437 Yellow Metal Lever-lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19338436 Supplier Eagle™ Supplier No. 1623BLK
Only null left

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Ideal for safe storage, handling or transportation of hazardous materials, spill containment, and bio waste disposal.

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids

Specifications

Product Type Drum
Closure Included Yes
Closure Type Plastic Lever-lock
Capacity (English) 20 gal.
Capacity (Metric) 75.8 L
Color Black
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Head Style Open
Height (English) 29.9 in.
Height (Metric) 75.9 cm
Weight (English) 10.25 lb.
Show More Show Less