missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lab Armor™ StayTemp™ Equipment Tray
For maintaining cold or heat with high efficiency in refrigerators, incubators, & ovens
Supplier: Lab Armor™ 39956003
Description
StayTemp Equipment trays are an upgrade that keeps samples and reagents in your refrigerator, incubator, and laboratory ovens at temperature despite normal hourly fluctuations in air temperature.
- Stainless steel construction
- Keeps samples organized protecting sample from defrost cycles and constant door opening and closing
- Eliminates the need of racks
- Can be hand washed, autoclaved or towel dried
- For any size and shape vessel including non-water-tight vessels
- Available in 3 sizes
Does not include lid.
Specifications
| 4 L | |
| 30.48 cm | |
| Equipment Tray | |
| For maintaining cold or heat with high efficiency in refrigerators, incubators and ovens | |
| 12 in. | |
| Rectangular | |
| 4 lb. | |
| 1.8 kg | |
| Autoclavable |
| 10.16 cm | |
| Stainless Steel | |
| 25.4 cm | |
| 4 L Lab Armor Beads | |
| 4 in. | |
| 356°F | |
| 180°C | |
| 10 in. |