missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L3MBTL1 Recombinant Human Monoclonal Antibody (RAB-C213)

L3MBTL1 Recombinant Human Monoclonal Antibody (RAB-C213)
Supplier: Fisher Chemical MA551864
Description
L3MBTL1 Recombinant Monoclonal Antibody for MS, ICC/IF, Flow, IP, ChIP, ELISA
Specifications
| L3MBTL1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q9Y468 | |
| L3MBTL1 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing L3MBTL1 protein under non-denaturing conditions. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| F(ab) κ |
| ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Immunocytochemistry | |
| RAB-C213 | |
| Unconjugated | |
| L3MBTL1 | |
| C630004G01; dJ138B7.3; H-l(3)mbt; H-l(3)mbt protein; Kiaa0681; L(3)mbt protein homolog; l(3)mbt-like (Drosophila); l(3)mbt-like 1; l(3)mbt-like 1 (Drosophila); L3mbt; L3MBTL; L3MBTL histone methyl-lysine binding protein 1; L3MBTL1; L3MBTL1, histone methyl-lysine binding protein; lethal (3) malignant brain tumor l(3); lethal(3)malignant brain tumor-like protein 1; lethal(3)malignant brain tumor-like protein 1; serine/threonine-protein kinase Sgk2; LOC100070683; mKIAA0681; RGD1307316; RP1-138B7.1; ZC2HC3 | |
| Human | |
| Metal-chelate Chromatography | |
| RUO | |
| 26013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |