missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Xylose, 99%

Catalog No. AAB2162206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2162206 5 g
AAB2162214 25 g
AAB2162222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2162206 Supplier Thermo Scientific Chemicals Supplier No. B2162206

CAS: 609-06-3 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-06-3
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00151096
InChI Key PYMYPHUHKUWMLA-UHFFFAOYNA-N
Synonym l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose
PubChem CID 95259
ChEBI CHEBI:17979
SMILES OCC(O)C(O)C(O)C=O

Specifications

Melting Point 147°C to 151°C
Density 1.525
Quantity 5 g
Beilstein 1723080
Sensitivity Hygroscopic
Optical Rotation −20° (c=10 in Water, 10h)
IUPAC Name (2S,3R,4S)-2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material L-(-)-Xylose
EINECSNumber 210-174-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only