missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Xylose, 99+%

CAS: 609-06-3 | C5H10O5 | 150.13 g/mol

Supplier:  Thermo Scientific Chemicals 225990050

 View more versions of this product

Catalog No. AC225990050


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(-)-Xylose
150.0°C to 152.0°C
0.5% max.
99% min. (ELSD) (HPLC)
C5H10O5
5 g
− 18.70 (24.00°C c=4,H2O,24h)
l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose
PYMYPHUHKUWMLA-UHFFFAOYNA-N
(2S,3R,4S)-2,3,4,5-tetrahydroxypentanal
95259
150.13
Crystalline Powder
609-06-3
White
Authentic
Glass bottle
MFCD00151096
31,55
− 18.70
Solubility in water: soluble. Other solubilities: soluble in pyridine and hot alcohol
OCC(O)C(O)C(O)C=O
150.13
CHEBI:17979
99+%

Chemical Identifiers

609-06-3
150.13
PYMYPHUHKUWMLA-UHFFFAOYNA-N
95259
OCC(O)C(O)C(O)C=O
C5H10O5
MFCD00151096
l-xylose, l +-xylose, l---xylose, 2s,3r,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-xylose, l-xylo-pentose, l-xyl, xylose, l, linear l-xylose, l--xylose
CHEBI:17979
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-174-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only