missing translation for 'onlineSavingsMsg'
Learn More

L-Valine 98.0+%, TCI America™

Catalog No. V001425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V001425G 25 g
V0014100G 100 g
V0014500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. V001425G Supplier TCI America Supplier No. V001425G
Only null left

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 117.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Valine
EINECSNumber (9)-1604
RTECSNumber YV9361000
TSCA Yes