missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ L-Valine methyl ester hydrochloride, 99%
$121.94 - $397.49
Chemical Identifiers
| CAS | 6306-52-1 |
|---|---|
| Molecular Formula | C6H13NO2·HCl |
| Molecular Weight (g/mol) | 167.64 |
| MDL Number | MFCD00012497 |
| InChI Key | KUGLDBMQKZTXPW-JEDNCBNOSA-N |
| Synonym | l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride |
| PubChem CID | 111190 |
| IUPAC Name | methyl (2S)-2-amino-3-methylbutanoate;hydrochloride |
| SMILES | CC(C)C(C(=O)OC)N.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC229260100
|
Thermo Scientific™
229260100 |
10g | Glass bottle |
Each for $121.94
|
|
||||
|
AC229260500
|
Thermo Scientific™
229260500 |
50g | Glass bottle |
Each for $397.49
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C6H13NO2·HCl | |
| MFCD00012497 | |
| l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride | |
| methyl (2S)-2-amino-3-methylbutanoate;hydrochloride |
Specifications
| 6306-52-1 | |
| 98.5% min. (Argentometry) | |
| (CH3)2CHCH(NH2)COOCH3·HCl | |
| +15.50° (24°C C=2,H2O) | |
| l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride | |
| KUGLDBMQKZTXPW-JEDNCBNOSA-N | |
| methyl (2S)-2-amino-3-methylbutanoate;hydrochloride | |
| 111190 | |
| 99% |
| Authentic | |
| C6H13NO2·HCl | |
| MFCD00012497 | |
| + 15.50 | |
| (5% in water) Clear colorless to light yellow | |
| CC(C)C(C(=O)OC)N.Cl | |
| 167.64 | |
| 167.64 | |
| L-Valine methyl ester hydrochloride, 99% |
Safety and Handling
EINECSNumber : 228-620-9
RUO – Research Use Only