missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Valine methyl ester hydrochloride, 99%

Catalog No. AC229260100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229260100 10 g Glass bottle
AC229260500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC229260100 Supplier Thermo Scientific Chemicals Supplier No. 229260100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6306-52-1 | C6H13ClNO2 | 166.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6306-52-1
Molecular Formula C6H13ClNO2
Molecular Weight (g/mol) 166.63
MDL Number MFCD00012497
InChI Key RFMIEHOYULVMNZ-UHFFFAOYNA-N
Synonym l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride
PubChem CID 111190
IUPAC Name methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
SMILES [Cl].COC(=O)C(N)C(C)C

Specifications

Melting Point 171.0°C to 173.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2CHCH(NH2)COOCH3·HCl
Quantity 10 g
Specific Rotation Condition +15.50° (24°C C=2,H2O)
Specific Rotation + 15.50
Solubility Information (5% in water) Clear colorless to light yellow
Formula Weight 167.64
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-Valine methyl ester hydrochloride, 99%
Show More Show Less
EINECSNumber 228-620-9

RUO – Research Use Only