missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ L-Valine methyl ester hydrochloride, 99%

Supplier:  Thermo Scientific™ 229260500

 View more versions of this product

Catalog No. AC229260500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

L-Valine methyl ester hydrochloride
6306-52-1
98.5% min. (Argentometry)
C6H13NO2·HCl
MFCD00012497
+15.50° (24°C C=2,H2O)
l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride
KUGLDBMQKZTXPW-JEDNCBNOSA-N
methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
111190
99%
99%
Authentic
Glass bottle
(CH3)2CHCH(NH2)COOCH3·HCl
50g
+ 15.50
(5% in water) Clear colorless to light yellow
CC(C)C(C(=O)OC)N.Cl
167.64
167.64

Chemical Identifiers

6306-52-1
167.64
KUGLDBMQKZTXPW-JEDNCBNOSA-N
111190
CC(C)C(C(=O)OC)N.Cl
C6H13NO2·HCl
MFCD00012497
l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride
methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 228-620-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only