missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine Ethyl Ester Hydrochloride 98.0+%, TCI America™
Supplier: TCI America V014225G
Specifications
| L-Valine Ethyl Ester Hydrochloride | |
| 104°C | |
| C7H16ClNO2 | |
| 25 g | |
| PQGVTLQEKCJXKF-RGMNGODLSA-N | |
| ethyl (2S)-2-amino-3-methylbutanoate hydrochloride | |
| 87181 | |
| ≥98.0% (T) |
| 17609-47-1 | |
| White | |
| MFCD00012511 | |
| l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl | |
| Cl.CCOC(=O)[C@@H](N)C(C)C | |
| 181.66 | |
| 181.66 | |
| Crystalline Powder |
Chemical Identifiers
| 17609-47-1 | |
| 181.66 | |
| PQGVTLQEKCJXKF-RGMNGODLSA-N | |
| 87181 | |
| Cl.CCOC(=O)[C@@H](N)C(C)C |
| C7H16ClNO2 | |
| MFCD00012511 | |
| l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl | |
| ethyl (2S)-2-amino-3-methylbutanoate hydrochloride |
Safety and Handling
TSCA : No