missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine Ethyl Ester Hydrochloride 98.0+%, TCI America™
$44.28 - $102.03
Chemical Identifiers
| CAS | 17609-47-1 |
|---|---|
| Molecular Formula | C7H16ClNO2 |
| Molecular Weight (g/mol) | 181.66 |
| MDL Number | MFCD00012511 |
| InChI Key | PQGVTLQEKCJXKF-RGMNGODLSA-N |
| Synonym | l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl |
| PubChem CID | 87181 |
| IUPAC Name | ethyl (2S)-2-amino-3-methylbutanoate hydrochloride |
| SMILES | Cl.CCOC(=O)[C@@H](N)C(C)C |
Chemical Identifiers
| 17609-47-1 | |
| 181.66 | |
| PQGVTLQEKCJXKF-RGMNGODLSA-N | |
| 87181 | |
| Cl.CCOC(=O)[C@@H](N)C(C)C |
| C7H16ClNO2 | |
| MFCD00012511 | |
| l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl | |
| ethyl (2S)-2-amino-3-methylbutanoate hydrochloride |
Specifications
| 17609-47-1 | |
| White | |
| MFCD00012511 | |
| l-valine ethyl ester hydrochloride, h-val-oet.hcl, ethyl l-valinate hydrochloride, h-val-oet hcl, valine ethyl ester hydrochloride, h-l-meval-ome*hcl, valine, ethyl ester, hydrochloride, ethyl 2s-2-amino-3-methylbutanoate hydrochloride, l-ethyl 2-amino-3-methylbutanoate hydrochloride, ethyl l-valinate hcl | |
| Cl.CCOC(=O)[C@@H](N)C(C)C | |
| 181.66 | |
| 181.66 | |
| Crystalline Powder |
| 104°C | |
| C7H16ClNO2 | |
| 5 g | |
| PQGVTLQEKCJXKF-RGMNGODLSA-N | |
| ethyl (2S)-2-amino-3-methylbutanoate hydrochloride | |
| 87181 | |
| ≥98.0% (T) | |
| L-Valine Ethyl Ester Hydrochloride |
Safety and Handling
TSCA : No