missing translation for 'onlineSavingsMsg'
Learn More

L-Valine, Cell Culture Reagent

Catalog No. p-7042559
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6294306 5 g
AAJ6294314 25 g
AAJ6294322 100 g
AAJ62943A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6294306 Supplier Thermo Scientific Chemicals Supplier No. J6294306
Only null left

CAS: 72-18-4 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Specifications

Color White
Beilstein 1721136
Merck Index 14,9909
Quantity 5 g
Odor Odorless
Formula Weight 117.15
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Valine
EINECSNumber 200-773-6
RTECSNumber YV9361000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only