missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine benzyl ester hydrochloride, 95%
CAS: 2462-34-2 | C12H18ClNO2 | 243.73 g/mol
$100.81 - $388.23
Chemical Identifiers
| CAS | 2462-34-2 |
|---|---|
| Molecular Formula | C12H18ClNO2 |
| Molecular Weight (g/mol) | 243.73 |
| MDL Number | MFCD22422648 |
| InChI Key | ZIUNABFUHGBCMF-MERQFXBCSA-N |
| Synonym | l-valine benzyl ester hydrochloride, val-obzl hcl, h-val-obzl.hcl, s-benzyl 2-amino-3-methylbutanoate hydrochloride, valine benzyl ester hydrochloride, benzyl 2s-2-amino-3-methylbutanoate hydrochloride, val-obzl.hcl, val-obzl. hcl, l-valine benzyl ester hydrochloride |
| PubChem CID | 11601206 |
| IUPAC Name | benzyl (2S)-2-amino-3-methylbutanoate;hydrochloride |
| SMILES | Cl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6224206
|
Thermo Scientific Chemicals
H6224206 |
5 g |
Each for $100.81
|
|
|||||
|
AAH6224214
|
Thermo Scientific Chemicals
H6224214 |
25 g |
Each for $388.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2462-34-2 | |
| 243.73 | |
| ZIUNABFUHGBCMF-MERQFXBCSA-N | |
| 11601206 | |
| Cl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1 |
| C12H18ClNO2 | |
| MFCD22422648 | |
| l-valine benzyl ester hydrochloride, val-obzl hcl, h-val-obzl.hcl, s-benzyl 2-amino-3-methylbutanoate hydrochloride, valine benzyl ester hydrochloride, benzyl 2s-2-amino-3-methylbutanoate hydrochloride, val-obzl.hcl, val-obzl. hcl, l-valine benzyl ester hydrochloride | |
| benzyl (2S)-2-amino-3-methylbutanoate;hydrochloride |
Specifications
| 138°C | |
| C12H18ClNO2 | |
| l-valine benzyl ester hydrochloride, val-obzl hcl, h-val-obzl.hcl, s-benzyl 2-amino-3-methylbutanoate hydrochloride, valine benzyl ester hydrochloride, benzyl 2s-2-amino-3-methylbutanoate hydrochloride, val-obzl.hcl, val-obzl. hcl, l-valine benzyl ester hydrochloride | |
| Cl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 11601206 | |
| 243.73 | |
| L-Valine benzyl ester hydrochloride |
| 2462-34-2 | |
| MFCD22422648 | |
| ZIUNABFUHGBCMF-MERQFXBCSA-N | |
| benzyl (2S)-2-amino-3-methylbutanoate;hydrochloride | |
| 243.73 | |
| −10° (c=2 in Water) | |
| 95% |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only