accessibility menu, dialog, popup

UserName

CAS RN 2462-34-2

H3CCH3(S)OONH2HCl

CAS RN 2462-34-2

IUPAC Name: benzyl (2S)-2-amino-3-methylbutanoate hydrochloride
Synonyms: benzyl (2S)-2-amino-3-methylbutanoate hydrochloride
Molecular Weight (g/mol): 243.73
Molecular Formula: C12H18ClNO2
InChi Key: ZIUNABFUHGBCMF-MERQFXBCSA-N
SMILES: Cl.CC(C)[C@H](N)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)