missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Valine, 99%

CAS: 72-18-4 | C5H11NO2 | 117.15 g/mol

Supplier:  Thermo Scientific Chemicals A1272022

 View more versions of this product

Catalog No. AAA1272022


Only null left

Description

Description

L-Valine is used in the enhancement of muscle growth and energy. It is utilized in tissue repair. It is also involved in the studies to attenuate arrhythmias and induce hypotensive effects. It acts as a precursor for the preparation of penicillin through biosynthetic pathway. It also employed as nutritional supplements in food and beverage industries. Further, it plays an important role in maintaining mental vigor, muscle coordination and emotional calm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Valine is used in the enhancement of muscle growth and energy. It is utilized in tissue repair. It is also involved in the studies to attenuate arrhythmias and induce hypotensive effects. It acts as a precursor for the preparation of penicillin through biosynthetic pathway. It also employed as nutritional supplements in food and beverage industries. Further, it plays an important role in maintaining mental vigor, muscle coordination and emotional calm.

Solubility
Soluble in water. Slightly soluble in methanol and acetone.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

∼315°C
72-18-4
C5H11NO2
1721136
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
KZSNJWFQEVHDMF-BYPYZUCNSA-N
CC(C)[C@H](N)C(O)=O
117.15
6287
Odorless
117.15
L-Valine
99%
MFCD00064220
14,9909
Soluble in water. Slightly soluble in methanol and acetone.
1.23
(2S)-2-amino-3-methylbutanoic acid
100 g
CHEBI:16414
+27° (c=5 in 5N HCl)
99%

Chemical Identifiers

72-18-4
117.15
KZSNJWFQEVHDMF-BYPYZUCNSA-N
6287
(2S)-2-amino-3-methylbutanoic acid
C5H11NO2
MFCD00064220
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
CHEBI:16414
CC(C)[C@H](N)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-773-6

RTECSNumber : YV9361000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only