missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Valine, 99%

Numéro de catalogue. AAA1272014
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1272014 25 g
AAA1272022 100 g
AAA1272036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AAA1272014 Fournisseur Thermo Scientific Chemicals Code fournisseur A1272014
Il en reste null

CAS: 72-18-4 | C5H11NO2 | 117.15 g/mol

L-Valine is used in the enhancement of muscle growth and energy. It is utilized in tissue repair. It is also involved in the studies to attenuate arrhythmias and induce hypotensive effects. It acts as a precursor for the preparation of penicillin through biosynthetic pathway. It also employed as nutritional supplements in food and beverage industries. Further, it plays an important role in maintaining mental vigor, muscle coordination and emotional calm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in methanol and acetone.

Notes
Incompatible with strong oxidizing agents.

Identifiants chimiques

CAS 72-18-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064220
InChI Key KZSNJWFQEVHDMF-BYPYZUCNSA-N
Synonym l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin
PubChem CID 6287
ChEBI CHEBI:16414
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(O)=O

Spécifications

Melting Point ∽315°C
Assay Percent Range 99%
Beilstein 1721136
Merck Index 14,9909
Solubility Information Soluble in water. Slightly soluble in methanol and acetone.
Density 1.23
Quantity 25 g
Formula Weight 117.15
Percent Purity 99%
Chemical Name or Material L-Valine
EINECSNumber 200-773-6
RTECSNumber YV9361000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only