missing translation for 'onlineSavingsMsg'
Learn More

L-Valinamide Hydrochloride 98.0+%, TCI America™

Catalog No. v01025g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01025G 5 g
V010225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01025G Supplier TCI America Supplier No. V01025G
Only null left

Chemical Identifiers

CAS 3014-80-0
Molecular Formula C5H13ClN2O
Molecular Weight (g/mol) 152.62
MDL Number MFCD00039085
InChI Key XFCNYSGKNAWXFL-WCCKRBBISA-N
Synonym l-valinamide hydrochloride, h-val-nh2 hcl, h-val-nh2.hcl, l-valine amide hydrochloride, 2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, l-valine amide, h-d-val-nh2 hcl, d-valinamide-hcl
PubChem CID 13689583
IUPAC Name (2S)-2-amino-3-methylbutanamide hydrochloride
SMILES Cl.CC(C)[C@H](N)C(N)=O

Specifications

Melting Point 263°C
Color White
Quantity 5 g
Formula Weight 152.62
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Valinamide Hydrochloride