accessibility menu, dialog, popup

UserName

H3CCH3(S)ONH2NH2HCl

CAS RN 3014-80-0

IUPAC Name: (2S)-2-amino-3-methylbutanamide hydrochloride
Synonyms: valinamide hydrochloride
Molecular Weight (g/mol): 152.62
Molecular Formula: C5H13ClN2O
InChi Key: XFCNYSGKNAWXFL-WCCKRBBISA-N
SMILES: Cl.CC(C)[C@H](N)C(N)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)