missing translation for 'onlineSavingsMsg'
Learn More

L-Valinamide hydrochloride, 95%

CAS: 3014-80-0 | C5H13ClN2O | 152.62 g/mol

Chemical Identifiers

CAS 3014-80-0
Molecular Formula C5H13ClN2O
Molecular Weight (g/mol) 152.62
MDL Number MFCD00039085
InChI Key XFCNYSGKNAWXFL-WCCKRBBISA-N
Synonym l-valinamide hydrochloride, h-val-nh2 hcl, h-val-nh2.hcl, l-valine amide hydrochloride, 2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, l-valine amide, h-d-val-nh2 hcl, d-valinamide-hcl
PubChem CID 13689583
IUPAC Name (2S)-2-amino-3-methylbutanamide;hydrochloride
SMILES Cl.CC(C)[C@H](N)C(N)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH6377506
View Documents Promotion Details
N/A 5 g N/A
Only null left
 
AAH6377514
View Documents Promotion Details
N/A 25 g N/A
Only null left
 
AAH6377522
View Documents Promotion Details
N/A 100 g N/A
Only null left
 
Description

Description

It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity

Solubility
Soluble in methanol (50 mg/ ml-clear, colorless solution).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

3014-80-0
152.62
XFCNYSGKNAWXFL-WCCKRBBISA-N
13689583
Cl.CC(C)[C@H](N)C(N)=O
C5H13ClN2O
MFCD00039085
l-valinamide hydrochloride, h-val-nh2 hcl, h-val-nh2.hcl, l-valine amide hydrochloride, 2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, l-valine amide, h-d-val-nh2 hcl, d-valinamide-hcl
(2S)-2-amino-3-methylbutanamide;hydrochloride

Specifications

266°C to 270°C
C5H13ClN2O
4198583
Soluble in methanol (50mg/ ml-clear,colorless solution).
Cl.CC(C)[C@H](N)C(N)=O
152.62
152.62
Solid
3014-80-0
MFCD00039085
l-valinamide hydrochloride, h-val-nh2 hcl, h-val-nh2.hcl, l-valine amide hydrochloride, 2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, l-valine amide, h-d-val-nh2 hcl, d-valinamide-hcl
XFCNYSGKNAWXFL-WCCKRBBISA-N
(2S)-2-amino-3-methylbutanamide;hydrochloride
13689583
95%
L-Valinamide hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only