missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine tert-Butyl Ester 97.0+%, TCI America™

Catalog No. T24465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24465G 5 g
T244625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24465G Supplier TCI America Supplier No. T24465G
Only null left

Chemical Identifiers

CAS 16874-12-7
Molecular Formula C13H19NO3
Molecular Weight (g/mol) 237.30
MDL Number MFCD00042644
InChI Key DIGHFXIWRPMGSA-LDGXTIHJNA-N
Synonym l-tyrosine tert-butyl ester, h-tyr-otbu, tert-butyl l-tyrosinate, l-tyrosine t-butyl ester, tert-butyl 2s-2-amino-3-4-hydroxyphenyl propanoate, h-tyr-obut, tyrosine, 1,1-dimethylethyl ester, 2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester, l-tyrosine tert.butyl ester, pubchem7068
PubChem CID 6950582
IUPAC Name tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES CC(C)(C)OC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 5 g
Formula Weight 237.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Tyrosine tert-Butyl Ester