accessibility menu, dialog, popup

UserName

(S)ONH2OHOCH3CH3CH3

CAS RN 16874-12-7

IUPAC Name: tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Synonyms: tert-butyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 237.3
Molecular Formula: C13H19NO3
InChi Key: DIGHFXIWRPMGSA-LDGXTIHJNA-N
SMILES: CC(C)(C)OC(=O)[C@@H](N)CC1=CC=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results