missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Methyl Ester 98.0+%, TCI America™

Supplier:  TCI America T273625G

 View more versions of this product

Catalog No. T273625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

L-Tyrosine Methyl Ester
135°C
C10H13NO3
25 g
MWZPENIJLUWBSY-VIFPVBQESA-N
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
70652
195.22
Crystalline Powder
1080-06-4
White-Yellow
MFCD00002392
l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
COC(=O)[C@@H](N)CC1=CC=C(O)C=C1
195.22
CHEBI:17215
≥98.0% (GC,T)

Chemical Identifiers

1080-06-4
195.22
MWZPENIJLUWBSY-VIFPVBQESA-N
70652
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
C10H13NO3
MFCD00002392
l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
CHEBI:17215
COC(=O)[C@@H](N)CC1=CC=C(O)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4490

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions