accessibility menu, dialog, popup

UserName

CAS RN 1080-06-4

IUPAC Name: methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Synonyms: tyrosine methyl ester|Tyr-OMe|methyl L-tyrosinate
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: MWZPENIJLUWBSY-VIFPVBQESA-N
SMILES: COC(=O)[C@@H](N)CC1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)