missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Methyl Ester 98.0+%, TCI America™

Catalog No. T27365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27365G 5 g
T273625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27365G Supplier TCI America Supplier No. T27365G

Chemical Identifiers

CAS 1080-06-4
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002392
InChI Key MWZPENIJLUWBSY-VIFPVBQESA-N
Synonym l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
PubChem CID 70652
ChEBI CHEBI:17215
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 5 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Tyrosine Methyl Ester
EINECSNumber (3)-4490
TSCA No
Recommended Storage Refrigerator