missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine methyl ester, 98%

CAS: 1080-06-4 | C10H13NO3 | 195.22 g/mol

$103.69 - $103.69

Chemical Identifiers

CAS 1080-06-4
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002392
InChI Key MWZPENIJLUWBSY-VIFPVBQESA-N
Synonym l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
PubChem CID 70652
ChEBI CHEBI:17215
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)[C@@H](N)CC1=CC=C(O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC140650050
SDS View Documents
Thermo Scientific Chemicals
140650050
5 g Glass bottle
Each for $103.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1080-06-4
195.22
MWZPENIJLUWBSY-VIFPVBQESA-N
70652
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
C10H13NO3
MFCD00002392
l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
CHEBI:17215
COC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

1080-06-4
White to Yellow
97.5% min. (HPLC)
C10H13NO3
MFCD00002392
14, 612
+ 25
Solubility in water: .. Other solubilities: 5% in methanol
COC(=O)[C@@H](N)CC1=CC=C(O)C=C1
195.22
CHEBI:17215
98%
L-Tyrosine methyl ester, 98%
133°C to 135°C
Authentic
Glass bottle
4-(HO)C6H4CH2CH(NH2)CO2CH3
5 g
+ 25.00 (20.00°C c=2.4,CH3OH)
l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
MWZPENIJLUWBSY-VIFPVBQESA-N
methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
70652
195.22
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-095-3

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only