missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine methyl ester, 98%

Catalog No. AC140650050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC140650050 5 g Glass bottle
AC140650250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC140650050 Supplier Thermo Scientific Chemicals Supplier No. 140650050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1080-06-4 | C10H13NO3 | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1080-06-4
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002392
InChI Key MWZPENIJLUWBSY-VIFPVBQESA-N
Synonym l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
PubChem CID 70652
ChEBI CHEBI:17215
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 133°C to 135°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2CH3
Quantity 5 g
Beilstein 14, 612
Specific Rotation Condition + 25.00 (20.00°C c=2.4,CH3OH)
Specific Rotation + 25
Solubility Information Solubility in water: .. Other solubilities: 5% in methanol
Formula Weight 195.22
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Tyrosine methyl ester, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-095-3

RUO – Research Use Only