missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine Hydrazide 98.0+%, TCI America™
Supplier: TCI America T182025G
Specifications
| L-Tyrosine Hydrazide | |
| White-Yellow | |
| MFCD00007609 | |
| l-tyrosine hydrazide, s-2-amino-3-4-hydroxyphenyl propanehydrazide, tyrosine, hydrazide, l, l-tyrosinehydrazide, l-tyrosine, hydrazide, 2s-2-amino-3-4-hydroxyphenyl propanehydrazide, l-tyrosinohydrazide, tyrosine hydrazide, tyrosine hydrazone | |
| NNC(=O)[C@@H](N)CC1=CC=C(O)C=C1 | |
| 195.22 | |
| 195.22 | |
| Crystalline Powder |
| 7662-51-3 | |
| C9H13N3O2 | |
| 25 g | |
| MWIXENPCUPDSOS-QMMMGPOBSA-N | |
| (2S)-2-amino-3-(4-hydroxyphenyl)propanehydrazide | |
| 72779 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 7662-51-3 | |
| 195.22 | |
| MWIXENPCUPDSOS-QMMMGPOBSA-N | |
| 72779 | |
| NNC(=O)[C@@H](N)CC1=CC=C(O)C=C1 |
| C9H13N3O2 | |
| MFCD00007609 | |
| l-tyrosine hydrazide, s-2-amino-3-4-hydroxyphenyl propanehydrazide, tyrosine, hydrazide, l, l-tyrosinehydrazide, l-tyrosine, hydrazide, 2s-2-amino-3-4-hydroxyphenyl propanehydrazide, l-tyrosinohydrazide, tyrosine hydrazide, tyrosine hydrazone | |
| (2S)-2-amino-3-(4-hydroxyphenyl)propanehydrazide |
Safety and Handling
TSCA : Yes