CAS RN 7662-51-3
CAS RN 7662-51-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Tyrosine Hydrazide 98.0+%, TCI America™
CAS: 7662-51-3 Molecular Formula: C9H13N3O2 Molecular Weight (g/mol): 195.22 MDL Number: MFCD00007609 InChI Key: MWIXENPCUPDSOS-QMMMGPOBSA-N Synonym: l-tyrosine hydrazide,s-2-amino-3-4-hydroxyphenyl propanehydrazide,tyrosine, hydrazide, l,l-tyrosinehydrazide,l-tyrosine, hydrazide,2s-2-amino-3-4-hydroxyphenyl propanehydrazide,l-tyrosinohydrazide,tyrosine hydrazide,tyrosine hydrazone PubChem CID: 72779 IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanehydrazide SMILES: NNC(=O)[C@@H](N)CC1=CC=C(O)C=C1