missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine Disodium Salt, MP Biomedicals
$101.19 - $291.75
Chemical Identifiers
| CAS | 122666-87-9 |
|---|---|
| Molecular Formula | C9H13NNa2O5 |
| Molecular Weight (g/mol) | 261.19 |
| MDL Number | MFCD03840558 |
| InChI Key | WFTCFVUQORELJZ-CMIWSEPMNA-L |
| Synonym | tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate |
| PubChem CID | 71587456 |
| IUPAC Name | disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate dihydrate |
| SMILES | O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10557325
|
MP Biomedicals Inc
0210557325 |
25 g |
Each for $101.19
|
|
|||||
|
ICN10557380
|
MP Biomedicals Inc
0210557380 |
100 g |
Each for $291.75
|
|
|||||
Chemical Identifiers
| 122666-87-9 | |
| 261.19 | |
| WFTCFVUQORELJZ-CMIWSEPMNA-L | |
| 71587456 | |
| O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O |
| C9H13NNa2O5 | |
| MFCD03840558 | |
| tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate | |
| disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate dihydrate |
Specifications
| 122666-87-9 | |
| MFCD03840558 | |
| WFTCFVUQORELJZ-CMIWSEPMNA-L | |
| disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate dihydrate | |
| 261.19 | |
| 225.2 |
| C9H13NNa2O5 | |
| tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate | |
| O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O | |
| 25 g | |
| 71587456 | |
| L-Tyrosine Disodium Salt |
Safety and Handling
Recommended Storage : 0 to 5°C