missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Disodium Salt, MP Biomedicals

Catalog No. icn10557325
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10557325 25 g
ICN10557380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10557325 Supplier MP Biomedicals Inc Supplier No. 0210557325

Chemical Identifiers

CAS 122666-87-9
Molecular Formula C9H13NNa2O5
Molecular Weight (g/mol) 261.19
MDL Number MFCD03840558
InChI Key WFTCFVUQORELJZ-CMIWSEPMNA-L
Synonym tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate
PubChem CID 71587456
IUPAC Name disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate dihydrate
SMILES O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O

Specifications

Quantity 25 g
Formula Weight 225.2
Chemical Name or Material L-Tyrosine Disodium Salt
Recommended Storage 0 to 5°C